Anti-TRP1 Rabbit Polyclonal Antibody
ANTIA250260-100
New Product
- Antibody type:Primary
- Antigen name:5,6 dihydroxyindole 2 carboxylic acid oxidase
- Antigen symbol:TRP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# P17643
- Antigen synonyms:CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|Glycoprotein 75|b-PROTEIN|DHICA oxidase
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:75 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human TYRP1 protein
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to TRP1 for IHC-P with samples derived from Human and Mouse.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 5 to 10 µg/ml
Type: Primary
Antigen: TRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse