Order Entry
Germany
ContactUsLinkComponent
Anti-TRP1 Rabbit Recombinant Antibody [clone: TYRP1/1564R]
Anti-TRP1 Rabbit Recombinant Antibody [clone: TYRP1/1564R]
Catalog # ANTIA250258-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-TRP1 Rabbit Recombinant Antibody [clone: TYRP1/1564R]
Catalog # ANTIA250258-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5,6 dihydroxyindole 2 carboxylic acid oxidase
  • Antigen symbol:
    TRP1
  • Clonality:
    Recombinant
  • Clone:
    TYRP1/1564R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17643
  • Antigen synonyms:
    CAS2|Catalase B|6-dihydroxyindole-2-carboxylic acid oxidase|GP75|CATB|Glycoprotein 75|b-PROTEIN|DHICA oxidase
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    75 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human TYRP1 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [TYRP1/1564R] antibody to TRP1 for IHC-P with samples derived from Human and Mouse.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: TRP1
Clonality: Recombinant
Clone: TYRP1/1564R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse