Order Entry
Germany
ContactUsLinkComponent
 
Anti-CREBBP Rabbit Polyclonal Antibody
Catalog # ANTIA11437-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CREBBP Rabbit Polyclonal Antibody
Catalog # ANTIA11437-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CBP
  • Antigen symbol:
    CREBBP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q92793
  • Antigen synonyms:
    KAT3A/|CREB binding protein|RTS|_HUMAN|RSTS|Crebbp|Cyclic AMP responsive enhancer binding protein|KAT3A|CREB-binding protein
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,05% Proclin 300
  • Molecular weight:
    300 kDa
  • Sequence:
    QASQGQAQVMNGSLGAAGRGRGAGMPYPTPAMQGASSSVLAETLTQVSPQMTGHAGLNTAQAGGMAKMGITGNTSPFGQPFSQAGGQPMGATGVNPQLASK
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 200 - 300 of human CREBBP (NP_004371.2)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to CREBBP for WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200

Type: Primary
Antigen: CREBBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat