Anti-Albumin Rabbit Recombinant Antibody [clone: ALB/6409R]
Catalog # ANTIA248505-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Growth inhibiting protein 20
- Antigen symbol:Albumin
- Clonality:Recombinant
- Clone:ALB/6409R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P02768
- Antigen synonyms:GIG20|DKFZp779N1935|ALBU_HUMAN|alb|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Albumin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:66 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ALB protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [ALB/6409R] antibody to Albumin for ELISA, WB and IHC-P with samples derived from human.
- Validated applications: ELISA, WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Albumin
Clonality: Recombinant
Clone: ALB/6409R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Human