Order Entry
Germany
ContactUsLinkComponent
Anti-Albumin Mouse Recombinant Antibody [clone: rALB/6412]
Anti-Albumin Mouse Recombinant Antibody [clone: rALB/6412]
Catalog # ANTIA248496-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Albumin Mouse Recombinant Antibody [clone: rALB/6412]
Catalog # ANTIA248496-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    alb
  • Antigen symbol:
    Albumin
  • Clonality:
    Recombinant
  • Clone:
    rALB/6412
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02768
  • Antigen synonyms:
    GIG20|DKFZp779N1935|ALBU_HUMAN|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Growth inhibiting protein 20|Albumin
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    66 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human ALB protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rALB/6412] antibody to Albumin for ELISA and IHC-P with samples derived from human.

  • Validated applications: ELISA, IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Albumin
Clonality: Recombinant
Clone: rALB/6412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human