Auftragserfassung
Germany
AuftragsverfolgungKontakt
Mouse Creatine Kinase B Type ELISA Kit | Avantor

Mouse Creatine Kinase B Type ELISA Kit

Mouse creatine kinase B type ELISA kit, 96 Tests
Catalog # ANTIA312157-96
Supplier:   Antibodies.com
Mouse Creatine Kinase B Type ELISA Kit
Catalog # ANTIA312157-96
Supplier:  Antibodies.com
CAS Number:  
Mouse Creatine Kinase B Type ELISA Kit
Mouse Creatine Kinase B Type ELISA Kit

Mouse creatine kinase B type ELISA kit, 96 Tests

Wechseln zu:
CAS Number:  
Price
565,00 €
Anmelden für die Verfügbarkeit
1 * 96 Tests
Product Details & Documents

Mouse Creatine Kinase B Type ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Creatine Kinase B Type in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 5 to 80 mU/ml
  • Sensitivity: 0,232 mU/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Creatine Kinase B Type ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Creatine Kinase B Type in serum, plasma or other biological fluids. An antibody specific for Creatine Kinase B Type has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Creatine Kinase B Type present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Creatine Kinase B Type antibody, which also binds the Creatine Kinase B Type present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Creatine Kinase B Type antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Creatine Kinase B Type antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Creatine Kinase B Type captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Creatine Kinase B Type in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Bestellinformationen: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Spezifikationen:

Spezifikationen:

  • VE:
    96 Tests
  • Bezeichnung:
    Mouse creatine kinase B type ELISA kit
  • Empfindlichkeit:
    0,232 mU/ml
  • Reaktivität der primären Antikörper:
    Mouse
  • Shelf life:
    Store for 6 months at 4 °C
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA312157-96
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Format:
    Pre-coated
  • Target protein:
    Creatine Kinase B Type
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erfassungsbereich:
    5 - 80 mU/ml
  • Regulatory status:
    RUO

Spezifikationen:

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.