- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Superoxide Dismutase 4/CCS
- Description:Human Superoxide Dismutase 4/CCS ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:150 - 2400 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:9,58 ng/L
- Regulatory status:RUO
Human Superoxide Dismutase 4/CCS ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Superoxide Dismutase 4/CCS in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 150 to 2400 ng/L
- Sensitivity: 9,58 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Superoxide Dismutase 4/CCS ELISA kit (A313665) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Superoxide Dismutase 4/CCS in An antibody specific for Superoxide Dismutase 4/CCS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Superoxide Dismutase 4/CCS present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Superoxide Dismutase 4/CCS antibody, which also binds the Superoxide Dismutase 4/CCS present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Superoxide Dismutase 4/CCS antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Superoxide Dismutase 4/CCS antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Superoxide Dismutase 4/CCS captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Superoxide Dismutase 4/CCS in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.