Rat PCNA ELISA Kit
Product Details & Documents
Rat PCNA ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PCNA in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat PCNA ELISA Kit (A79606) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat PCNA in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PCNA has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PCNA present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PCNA Antibody, which binds the captured PCNA present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PCNA captured in each well. The concentration of PCNA can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.
Bestellinformationen: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Spezifikationen:
Spezifikationen:
- Pk:96 Tests
- Description:Rat PCNA ELISA kit
- Primary antibody reactivity:Rat
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:100 μl
- Detection range:0,156 - 10 ng/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Rat PCNA ELISA Kit exhibits high specificity and excellent specificity for the detection of rat PCNA. No significant cross-reactivity or interference between PCNA and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:ANTIA79606-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0,094 ng/ml
- Format:Pre-coated
- Target protein:PCNA
- Regulatory status:RUO
Spezifikationen:
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



