Human Anti-SARS-CoV-2 (S) IgG ELISA Kit
Catalog # ANTIA303150-96
Supplier: Antibodies.com
New Product
CAS Number:
Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Anti-SARS-CoV-2 (S) IgG
- Description:Human anti-SARS-CoV-2 (S) IgG ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:3,906 - 250 ng/ml
- Storage temperature:4 °C
- Sensitivity:2,344 ng/ml
- Regulatory status:RUO
Specifications
About this item
Human Anti-SARS-CoV-2 (S) IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 (S) IgG in serum, plasma, tissue homogenates, and other biological fluids.