- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:beta 2 Adrenergic receptor
- Description:Mouse beta 2 adrenergic receptor ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Mouse beta 2 Adrenergic Receptor ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse beta 2 Adrenergic Receptor. No significant cross-reactivity or interference between beta 2 Adrenergic Receptor and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,156 - 10 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,094 ng/ml
- Regulatory status:RUO
Mouse beta 2 Adrenergic Receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse beta 2 Adrenergic Receptor in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample type: Serum, plasma, tissue homogenates and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse beta 2 Adrenergic Receptor ELISA kit (A76110) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse beta 2 Adrenergic Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for beta 2 Adrenergic Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the beta 2 Adrenergic Receptor present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-beta 2 Adrenergic Receptor Antibody, which binds the captured beta 2 Adrenergic Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of beta 2 Adrenergic Receptor captured in each well. The concentration of beta 2 Adrenergic Receptor can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.