DSPE-PEG-DBCO, M.W. 2000, Avanti Research™
Catalog # SIAL880229P-50MG
Supplier: Merck
New Product
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
Storage Temperature:Freezer
CAS Number:2052955-83-4
Formula:(C₂H₄O)nC₆₃H₁₀₀N₃O₁₂P.H₃N