Order Entry
Germany
ContactUsLinkComponent
Human SARS-CoV-2 Spike Protein Inhibitor Screening Kit
Human SARS-CoV-2 Spike Protein Inhibitor Screening Kit
Catalog # STMC100-0700
CAS Number:  
undefined
Human SARS-CoV-2 Spike Protein Inhibitor Screening Kit
Catalog # STMC100-0700
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    SARS-CoV-2

Specifications

About this item

For detection and measurement of compounds that inhibit binding of viral SARS-CoV-2 Spike protein to human ACE2 receptor.

The Human SARS-CoV-2 Spike Protein Inhibitor Screening Kit is designed for the identification and characterization of inhibitor molecules, including antibodies or chemicals, that prevent the binding of SARS-CoV-2 virus to the host angiotensin-converting enzyme 2 (ACE-2)-expressing cells. The Spike (S) protein is a type I transmembrane glycoprotein present on the surface of coronaviruses (CoV). Entry of CoV, including SARS-CoV-2, into host cells is mediated by the S protein, where it interacts with the cell-surface receptor ACE2. In humans, ACE2 is expressed in several organs and tissues, including the intestinal and respiratory epithelia.

The S protein has been shown to play a key role in the induction of neutralizing antibody and T cell responses, which may lead to protective immunity. This inhibitor screening assay is based on a competitive ELISA method, in which inhibitors to be tested are mixed with biotinylated recombinant human ACE2 and are added to ELISA strip plates coated with recombinant SARS-CoV-2 S protein.. The captured biotinylated ACE2 is detected by streptavidin-horseradish peroxidase (HRP), which binds the biotinylated ACE2.

Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a coloured product with an intensity directly proportional to the binding activity of ACE2 to the S protein. Inhibitors will compete with the biotinylated ACE2 for binding to the SARS-CoV-2 S protein, resulting in reduced signal for more effective inhibitors. This inhibitor screening assay can be performed in the reverse configuration, in which the plate is coated with biotinylated recombinant ACE2, recombinant S Protein is mixed with the inhibitor to be tested, and S protein binding is detected using anti-human IgG-HRP.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').