About this item
For detection and measurement of mouse interleukin 12 (p70) subunit.
The Mouse Interleukin 12 (IL-12) p70 ELISA Kit is designed for the quantitative detection and measurement of mouse IL-12 (p70) in biological fluids such as serum, plasma, and cell culture supernatants. IL-12 (p70) is a heterodimeric cytokine composed of p35 and p40 subunits; the p40 subunit is shared with IL-23. IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils, and B cells.
IL-12 is a regulator of cytokine synthesis, especially IFN-γ, enhances the proliferation of T and NK cells, and stimulates the differentiation of CD⁴⁺ and CD⁸⁺ T cells. The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine.
The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’, 5,5’ tetramethylbenzidine (TMB) results in a coloured product with an intensity directly proportional to the concentration of cytokine in the sample.
The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analysed in parallel.
Specifications
- Primary antibody reactivity:Mouse
- Target protein:Interleukin-12