Order Entry
Germany
ContactUsLinkComponent
3M™ Professional Masking Tape
3M™ Professional Masking Tape
Catalog # MMMA209036SW
Supplier:  3M
New Product
3M™ Professional Masking Tape
Catalog # MMMA209036SW
Supplier:  3M
New Product

Specifications

  • Width:
    36 mm
  • Colour:
    Blue
  • Description:
    3M™ professional masking tape
  • Length:
    50 m
  • Pk:
    1 Roll

Specifications

About this item

3M™ Professional Masking Tape 2090 is a universal tape that can be used to mask various surfaces such as glazing, glass, wood, plastic, tiles and metal.

  • UV and water resistant
  • Ideal for glass, wood, plastic, tiles and metal, indoors and outdoors
  • The masking tape is made from PEFC certified paper, whose forests are managed sustainably and responsibly

Use 3M™ Professional Masking Tape 2090 to achieve clean paint lines on all interior and exterior surfaces. It adheres evenly to most surfaces without bleed-through, ensuring perfect paint lines. It can be removed cleanly without residue for up to 14 days after application.

Adhesive type: Acrylic