ContactUsLinkComponent
Crimp Vial, Quartz Glass
Crimp Vial, Quartz Glass
  NEUB7820-QU-04
New Product
Crimp Vial, Quartz Glass
  NEUB7820-QU-04
New Product
  • Cap included:
    No
  • Ø×H:
    15×45 mm
  • Capacity:
    4,00 ml
  • Description:
    Crimp vials made of quartz glass
  • Material:
    Quartz
  • Pk:
    1
Crimp vial made of quartz glass.
  • Without cap
  • Without labeling