Order Entry
Germany
ContactUsLinkComponent
SureBlue™ TMB 1-Component Microwell Peroxidase Substrate, KPL
SureBlue™ TMB 1-Component Microwell Peroxidase Substrate, KPL
Catalog # KPLI5120-0075
CAS Number:  
SureBlue™ TMB 1-Component Microwell Peroxidase Substrate, KPL
Catalog # KPLI5120-0075
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    SureBlue™ TMB 1-Component microwell peroxidase substrate
  • Seq:
    01
  • Shelf life:
    36 months
  • Storage temperature:
    Store at 2...8 °C

Specifications

About this item

SureBlue™ TMB Microwell substrate develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm and turns yellow when stopped by acidification and read at 450 nm.

Recommended for ELISA. Single component, ready to use format of 3,3',5,5' - Tetramethylbenzidine. No reagent preparation required, very convenient.