LABMAX Premium Bottle-Top Dispensers
Supplier: witeg Labortechnik
New Product
Certificates
About this item
The universal dispenser LABMAX premium has a sophisticated system which allows air purging and draining without any loss of reagent. Due to the direct displacement system, LABMAX premium is ideal for crystalline solutions. We improved the adjust knob for a quick and precise volume setting. On top, we added the built-in recalibration mechanism.
- Easy to clean, completely autoclavable at 121 °C
- Highest accuracy and precision
- All-purpose dispenser, universal chemical compatibility
- Direct displacement piston system, ideal for crystalline solutions
- Built-in recalibration mechanism including calibration tool
- Robust and stable, made of high quality materials e.g. platinum springs
- One handed operation and flexible by using different bottle adapters
0% Loss of reagent by pumping in a closed circuit, 100% drip-free due to our unique anti-drip system. 360° rotatable valve block for added safety with 2 scales - for measurements in milliliter and ounces. Telescopic suction tube included in scope of supply.
Delivery: Supplied with telescopic suction tube, discharge tube, instruction manual, calibration tool, individual certificate, serial number, thread adapter.