Order Entry
Germany
ContactUsLinkComponent
Lithium hydroxide monohydrate ≥99.9% (trace metals basis) battery grade
Corrosive
Irritant
Lithium hydroxide monohydrate ≥99.9% (trace metals basis) battery grade
Catalog # SIAL930903-100G
Supplier:  Merck
New Product
CAS Number:  
Corrosive
Irritant
Lithium hydroxide monohydrate ≥99.9% (trace metals basis) battery grade
Catalog # SIAL930903-100G
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance (Colour):
    White
  • Appearance (Form):
    Powder
  • Titration with HCl
    %LiOH:
    ≥56.5 %
  • Chloride:
    ≤ 50 ppm
  • Sulfate:
    ≤ 50 ppm
  • ICP Major Analysis
    Confirms Lithium component:
    Confirmed
  • Purity
    >=99.9% Based on Trace Metals Analysis:
    Meets Requirements
  • Trace Metal Analysis:
    ≤1000.0 ppm

Specification Test Results

About this item

 :
 :41.96 g/mol
Melting Pt:423 °C
 :M-Clarity™ quality level = MQ100
 :GHS05
 :GHS07
CAS Number:1310-66-3
Formula:LiOH·1H₂O
MDL Number:MFCD00149772