Order Entry
Germany
ContactUsLinkComponent
AzoDye-3 succinimidyl ester
AzoDye-3 succinimidyl ester
Catalog # AATB2472
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
AzoDye-3 succinimidyl ester
Catalog # AATB2472
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :680.21 g/mol
Storage Temperature:Freezer