Order Entry
Germany
ContactUsLinkComponent
AzoDye-2-PEG10 azide
AzoDye-2-PEG10 azide
Catalog # AATB2433
Supplier:  AAT Bioquest
New Product
CAS Number:  
AzoDye-2-PEG10 azide
Catalog # AATB2433
Supplier:  AAT Bioquest
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1015.13 g/mol
Storage Temperature:Freezer