Order Entry
Germany
ContactUsLinkComponent
AzoDye-1 PEG10 succinimidyl ester
AzoDye-1 PEG10 succinimidyl ester
Catalog # AATB2403
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
AzoDye-1 PEG10 succinimidyl ester
Catalog # AATB2403
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1113.23 g/mol
Storage Temperature:Freezer