Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ferroptocide
Ferroptocide ≥90%
Ferroptocide ≥90%
Ferroptocide ≥90%
CAYM39005-5
:
Cayman Chemical
New Product
:
Ferroptocide ≥90%
CAYM39005-5
:
Cayman Chemical
New Product
:
Pk:
5 mg
Seq:
2210
:
:
586.08 g/mol
:
Room Temperature
:
2505218-38-0
:
C₃₀H₃₆ClN₃O₇