Magicard™ Prima 8 Duo Printers
: Brady
New Product
The Magicard Prima 8 printer provides a compact, reliable and robust solution for instant issuance and lamination.
- High quality double-sided colour card printing, even to the edges
- Equipped with a range of encoding devices to write secure electronic data onto cards at the point of issue
- Smartcard encoding enables printing and programing your smartcard in one go
- Application: Health and hospital labeling, facility identification, safety identification, general identification
Using a retransfer printing process, images are first printed onto a thin film and then thermally transferred to the card surface. This produces a high-resolution glossy image, printed to the edges of the card.
Visual security: Hologram, Uv-effect
- Catalog No:
- BRDY321954
- BRDY321955
- BRDY321956
- Compatibility:
- CR79, CR80
- CR79, CR80
- CR79, CR80
- Connections:
- Ethernet, USB
- Ethernet, USB
- Ethernet, USB
- Display:
- Monochrome LCD
- Monochrome LCD
- Monochrome LCD
- Electrical:
- 110 - 240 V
- 110 - 240 V
- 110 - 240 V
- Modes:
- Automatic, manual
- Automatic, manual
- Automatic, manual
- Print method:
- Thermal transfer, double-sided printing
- Thermal transfer, double-sided printing
- Thermal transfer, double-sided printing
- Print resolution:
- 600 dpi
- 600 dpi
- 600 dpi
- Print speed:
- Multi-colour print speed (per hour): 190
Single colour print speed (per hour): 750 - Monochrome print speed: 750 per hour
Colour printing speed: 190 per hour - Monochrome print speed: 750 per hour
Colour printing speed: 190 per hour
- Supported OS:
- Windows® 10, Windows® 7, Windows® 8, Windows® Server 2012, Windows® Server 2016
- Windows® 10, Windows® 7, Windows® 8, Windows® Server 2012, Windows® Server 2016
- Windows® 10, Windows® 7, Windows® 8, Windows® Server 2012, Windows® Server 2016
- Temperature range:
- 15...30 °C
- 15...30 °C
- 15...30 °C