About this item
Mouse monoclonal IgG2a antibody against mouse NK1.1 (CD161), PerCP-Cy5.5-conjugated.
- Nk cells
The PK136 antibody reacts with murine NK1.1 (CD161), an ~80 kDa homodimeric type 2 transmembrane glycoprotein and C-type lectin receptor expressed on NK cells, NK-T cells, and rare subsets of T cells and monocytes in select strains of mice. NK1.1 is encoded by the Klrb1b and Klrb1c genes, which specify CD161b and CD161c polypeptides, respectively. CD161b is expressed only by Swiss NIH and SJL mice, and CD161c by certain strains such as C57BL, FVB/N, and NZB (but not A, AKR, BALB/c, CBA/J, C3H, C57BR, C58, DBA/1, DBA/2, NOD, SJL, or 129). NK1.1 has functional roles in modulating several processes, including the activation and proliferation of NK cells, induction of interferon-γ production, and release of cytotoxic granules. Its expression on the cell surface is specifically upregulated by IL-12. For detection of NK cells in NK1.1- strains, Anti-Mouse CD49b Antibody, Clone DX5 (Catalog #60020) is recommended. DX5 recognizes CD49b (integrin α2), another commonly used NK cell marker.
Type:
Antigen: NK1.1 (CD161)
Clonality: Monoclonal
Clone: PK136
Conjugation: PerCP-Cy5.5
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Specifications
- Catalog No:
- STMC60103PS
- STMC60103PS.1
- Antigen name:
- NK1.1 (CD161)
- NK1.1 (CD161)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- PK136
- PK136
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 17059
- 17059
- Isotype:
- IgG2a kappa
- IgG2a kappa