About this item
Mouse monoclonal IgG2b antibody against human, rhesus, cynomolgus CD4, APC-conjugated.
- T cells
- T cells CD4
- T cells Regulatory
The OKT4 antibody reacts with CD4, an ~59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4). The epitope for OKT4 has been localized to the D3 domain of the protein, which has a structure resembling an Ig variable domain. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper cells), and at lower levels by peripheral blood monocytes and macrophages. CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the affinity of the association between the TCR and MHC II-antigen complex. CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck. Moreover, CD4 is a receptor for human immunodeficiency virus (HIV). Binding of the OKT4 antibody to CD4 does not block HIV binding. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD4+ T Cell Isolation Kit (Catalog #19662), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851) and EasySep™ Human CD4 Positive Selection Kit II (Catalog #17852).
Type:
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Chimpanzee;Cynomolgus monkey;Human;Rhesus monkey;Sooty mangabey
Specifications
- Catalog No:
- STMC60016AZ.1
- STMC60016AZ
- Antigen name:
- CD4
- CD4
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- OKT4
- OKT4
- Reactivity:
- Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Chimpanzee, Human
- Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 920
- 920
- Isotype:
- IgG2b kappa
- IgG2b kappa