Order Entry
Germany
ContactUsLinkComponent
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (Alexa Fluor® 488)
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (Alexa Fluor® 488)
  STMC60107AD
undefined
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (Alexa Fluor® 488)
  STMC60107AD

 

  • Antigen name:
    CD83
  • Clonality:
    Monoclonal
  • Clone:
    HB15e
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Rhesus monkey,
    Macaque,
    Chimpanzee,
    Human
  • Gene ID:
    9308
  • Pk:
    100 Tests

 

 

Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD83, Alexa Fluor® 488-conjugated.

  • Dendritic cells

The HB15e antibody reacts with CD83, an ~45 kDa type I transmembrane glycoprotein and member of the Siglec protein family, containing an extracellular V-type Ig-like domain and a C-terminal cytoplasmic tail. A soluble form of CD83 has also been identified. CD83 is expressed predominantly on circulating and tissue dendritic cells, including Langerhan cells and interdigitating reticulum cells. It is expressed weakly on activated lymphocytes and may also be found on neutrophils, monocytes and macrophages. Expression of CD83 is typically transient, except on dendritic cells, and correlates with upregulation of HLA class II antigen. Its function, though ill-defined, is essential for the development and maintenance of the immune system and evidence suggests it may be involved in cell adhesion and signaling, antigen presentation, and lymphocyte activation. The soluble form of CD83 appears to be immunosuppressive. The epitope on CD83 bound by the HB15e antibody reportedly overlaps with that of antibody 1E11.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Chimpanzee, Cynomolgus monkey, Human, Macaque, Rhesus monkey