Order Entry
Germany
ContactUsLinkComponent
 
Anti-CD45.1 Mouse Monoclonal Antibody [clone: A20] (Biotin)
 
undefined
Anti-CD45.1 Mouse Monoclonal Antibody [clone: A20] (Biotin)

 

  • Catalog No:
  • STMC60117BT
  • STMC60117BT.1
  • Antigen name:
  • CD45.1
  • CD45.1
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • A20
  • A20
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa