Order Entry
Germany
ContactUsLinkComponent
 
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (FITC)
 
undefined
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (FITC)

 

  • Catalog No:
  • STMC60082FI
  • STMC60082FI.1
  • Antigen name:
  • CD5
  • CD5
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • UCHT2
  • UCHT2
  • Reactivity:
  • Capuchin monkey, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human, Owl monkey
  • Capuchin monkey, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human, Owl monkey
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 921
  • 921
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa