Order Entry
Germany
ContactUsLinkComponent
Anti-F4/80 Rat Monoclonal Antibody [clone: BM8] (PerCP-Cy5.5)
Anti-F4/80 Rat Monoclonal Antibody [clone: BM8] (PerCP-Cy5.5)
Certificates
undefined
Anti-F4/80 Rat Monoclonal Antibody [clone: BM8] (PerCP-Cy5.5)

Specifications

  • Catalog No:
  • STMC60027PS
  • STMC60027PS.1
  • Antigen name:
  • F4/80
  • F4/80
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • BM8
  • BM8
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 13733
  • 13733
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications