Order Entry
Germany
ContactUsLinkComponent
Anti-Beta-tubulin III Mouse Monoclonal Antibody [clone: AA10] (Alexa Fluor® 488)
Anti-Beta-tubulin III Mouse Monoclonal Antibody [clone: AA10] (Alexa Fluor® 488)
Certificates
undefined
Anti-Beta-tubulin III Mouse Monoclonal Antibody [clone: AA10] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60100AD
  • STMC60100AD.1
  • Antigen name:
  • Beta-tubulin III
  • Beta-tubulin III
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • AA10
  • AA10
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 10381
  • 10381
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications