Methanol
1 mL
Catalog # CMPLSB243.1000
Supplier: CUSTOM MADE CHEMICALS LAB
Jump to:
Product Details & Documents
Ordering information: Shelf life on ship date: 24 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Boiling Pt:64,7 °C
- Melting Pt:–98 °C
- Density:0,79 g/cm³
- Flash Pt:11 °C
- EINECS:200-659-6
- UN:1230
- ADR:3 (6.1),II
Specifications
Specifications
- Pk:1 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



