SARS-CoV-2 Recombinant Nucleocapsid Protein, aa1-419 (HEK293-expressed)
Supplier: STEMCELL Technologies
Certificates
About this item
SARS-CoV-2 Recombinant Nucleocapsid Protein, aa1-419 is expressed in HEK293 cells and is one of four structural proteins encoded by the SARS-CoV-2 genome. The Nucleocapsid Protein is transcribed from the viral “N” gene and is the protein that interacts with RNA to form the nucleocapsid. The protein forms a homo-oligomer, and both the monomer and the oligomer can interact with RNA. This protein also interacts with the membrane protein (protein M) after infection of the host cell during packaging of the positive-strand viral genome RNA into the ribonucleocapsid during virion assembly.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:Nucleocapsid protein|NC|Protein N|N|N Protein
- Protein/peptide name:CoV-2 Recombinant Nucleocapsid Protein, aa1-419
- Purity:≥90%