Order Entry
Germany
ContactUsLinkComponent
Dulbecco's Phosphate Buffered Saline (D-PBS), 10X Concentrate without Ca++ and Mg++
Dulbecco's Phosphate Buffered Saline (D-PBS), 10X Concentrate without Ca++ and Mg++
Catalog # STMC37354
CAS Number:  
undefined
Dulbecco's Phosphate Buffered Saline (D-PBS), 10X Concentrate without Ca++ and Mg++
Catalog # STMC37354
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Dulbecco’s phosphate-buffered saline, 10X concentrate, without calcium and magnesium

Specifications

About this item

Dulbecco’s phosphate-buffered saline (D-PBS) can be used as a temporary diluting, washing, irrigating or transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media and provides cells with a source of water and essential inorganic ions.

Species: Human, mouse, non human primate, other, rat

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').