RoboSep™ Tip Head Polishing Compound
Catalog # STMC20119
Supplier: STEMCELL Technologies
Specifications
- Description:RoboSep™ support reagent
- Pk:7 mL
Specifications
About this item
The RoboSep™ Tip head polishing compound is a cleaning/polishing solution designed to remove residue build-up which may accumulate over time, thus improving the tip contact surface condition for pick up/stripping of disposable RoboSep™ Tips.
Application: Immunology
Species: Human, mouse, non human primate, other, rat
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').