Order Entry
Germany
Orders LinkContactUsLinkComponent
 

EasySep™ Red Blood Cell Lysis Buffer

Marketsource
EasySep™ Red Blood Cell Lysis Buffer
Marketsource
Certificates
EasySep™ Red Blood Cell Lysis Buffer
EasySep™ Red Blood Cell Lysis Buffer

2 Options Available Below

Product Details & Documents

EasySep™ Red blood cell lysis buffer is for use as directed with EasySep™ or RoboSep™ whole blood kits for the lysis of red blood cells prior to separation from whole blood.

Application: Chimerism, Hla, immunology
Species: Human

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • STMC20110
    Red blood cell lysis buffer, 10X concentrate
    Red blood cell lysis buffer, 10X concentrate
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 74,70 €
    74,70 €
    1 * 10 mL
       74,70 €

    Specifications:
    Marketsource
    Marketsource
  • STMC20120
    Red blood cell lysis buffer, 10X concentrate
    Red blood cell lysis buffer, 10X concentrate
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 695,00 €
    695,00 €
    1 * 100 mL
       695,00 €

    Specifications:
    Marketsource
    Marketsource