Human Recombinant EPO (CHO-expressed)
About this item
The principal physiological regulator of erythropoiesis, Erythropoietin (EPO) is a glycoprotein growth factor that is produced primarily in the kidney in response to hypoxia or anemia. EPO promotes erythropoiesis by binding to a homodimeric cell surface receptor that activates JAK2/STAT5, PI3K/AKT, and MAPK pathways, and stimulates the proliferation and differentiation of erythroid progenitor cells (Jelkmann. Transfus Med Hemother, 2013; Kuhrt and Wojchowski. Blood, 2015).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
3 Options Available Below
Product Details & Documents
The principal physiological regulator of erythropoiesis, Erythropoietin (EPO) is a glycoprotein growth factor that is produced primarily in the kidney in response to hypoxia or anemia. EPO promotes erythropoiesis by binding to a homodimeric cell surface receptor that activates JAK2/STAT5, PI3K/AKT, and MAPK pathways, and stimulates the proliferation and differentiation of erythroid progenitor cells (Jelkmann. Transfus Med Hemother, 2013; Kuhrt and Wojchowski. Blood, 2015).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- STMC78007.2
- STMC78007
- STMC78007.1
- Protein/peptide name:
- EPO
- EPO
- EPO
- Protein synonyms:
- EP|Epoetin|Erythropoietin
- EP|Epoetin|Erythropoietin
- EP|Epoetin|Erythropoietin
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Source:
- CHO cells
- CHO cells
- CHO cells
- Purity:
- ≥95%
- ≥95%
- ≥95%
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.STMC78007.2Protein/peptide nameEPOProtein synonymsEP|Epoetin|ErythropoietinProtein/peptide typeRecombinantSpeciesHumanSourceCHO cellsPurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78007Protein/peptide nameEPOProtein synonymsEP|Epoetin|ErythropoietinProtein/peptide typeRecombinantSpeciesHumanSourceCHO cellsPurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78007.1Protein/peptide nameEPOProtein synonymsEP|Epoetin|ErythropoietinProtein/peptide typeRecombinantSpeciesHumanSourceCHO cellsPurity≥95%Marketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



