Order Entry
Germany
ContactUsLinkComponent
Human Recombinant PDGF-CC
Human Recombinant PDGF-CC
Certificates
undefined
Human Recombinant PDGF-CC

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    PDGF-CC
  • Purity:
    ≥95%

Specifications