Order Entry
Germany
ContactUsLinkComponent
Human Recombinant IL-17A
Human Recombinant IL-17A
Certificates
undefined
Human Recombinant IL-17A

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IL-17A
  • Purity:
    ≥95%

Specifications