Order Entry
Germany
ContactUsLinkComponent
SARS-CoV-2 (Nucleocapsid Protein) Peptide Pool
SARS-CoV-2 (Nucleocapsid Protein) Peptide Pool
Catalog # STMC100-0647
CAS Number:  
undefined
SARS-CoV-2 (Nucleocapsid Protein) Peptide Pool
Catalog # STMC100-0647
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Protein/peptide name:
    SARS-CoV-2 (Nucleocapsid Protein) Peptide Pool
  • Purity:
    Average 70%

Specifications

About this item

The SARS-CoV-2 (Nucleocapsid protein) Peptide pool is a lyophilised mixture of 102 peptides from the nucleocapsid protein of SARS-CoV-2. Nucleocapsid protein is one of four structural proteins that interacts with RNA to form the nucleocapsid. It also interacts with membrane protein (protein M) in the packaging of positive-strand viral genome RNA during virion assembly. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 419 on SARS-CoV-2 nucleocapsid protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').