Gas Washing Bottle, Drechsel Pattern
NEUB05503000251906
New Product
- NS:18,8/26
- Capacity:25 ml
- Description:Gas washing bottle, after Drechsel, without frit
- Pk:1
The bottle is made up of borosilicate 3.3 glass.
Complete with NS 29/32 and DIN 12596, including a safety attachment. For bottles up to 2 liters, the base height is 200 mm. Bottles exceeding 2 liters have a taller base height.