Bovine Catalase (from Liver)
Catalog # ENZOSV-26910-01
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:250 mg
- CAS No.:9001-05-2
- Source:Liver
- Species:Bovine
- Storage conditions:−20 °C
- Shipping conditions:Ambient temperature
- Enzyme name:Catalase
- Enzyme activity:11000 U/mg
Specifications
About this item
Caution: Research use only.