Orders LinkContactUsLinkComponent
Copper-EDTA, Buffer Solution pH 10
Copper-EDTA, Buffer Solution pH 10
Copper EDTA buffer solution, 10, 5 L
  KRAF22908.5000
Copper-EDTA, Buffer Solution pH 10
  KRAF22908.5000
 :  
Copper-EDTA, Buffer Solution pH 10
Copper EDTA buffer solution, 10, 5 L
 :  
180,00 €
1 * 5 L
Copper-EDTA buffer solution pH 10 with 4,5 mM copper-EDTA for complexometric titration, METROHM, 5 L.
  • Pk:
    5 L
  • Description:
    Copper EDTA buffer solution
  • pH value:
    10