Biopsy Punch
Biopsy punch, sterile, 6 mm
Catalog # MEGRKAI601
Supplier: MEGRO
Jump to:
Product Details & Documents
Biopsy punch for precise skin biopsies with minimal tissue trauma.
- Ribbed handle for secure grip
- Made from specially hardened rustproof steel
Handle: Polypropylene
Packaging: Individually packaged.
Specifications
- Description:Biopsy punch, sterile, 6 mm