Order Entry
Germany
ContactUsLinkComponent
ESD Red Elasticated Adjustable Wrist Band
ESD Red Elasticated Adjustable Wrist Band
Catalog # DESC229975
Supplier:  DESCO
New Product
ESD Red Elasticated Adjustable Wrist Band
Catalog # DESC229975
Supplier:  DESCO
New Product

Specifications

  • Colour:
    Red
  • Description:
    Adjustable elastic wristband only, 4 mm
  • Pk:
    100

Specifications

About this item

Designed to be used with a coiled cord (included with wrist straps) with a one megohm current limiting resistor meeting the EN 61340-5-1.

  • No metal in skin contact, no skin allergies
  • Circumference: adjustable from 120 to 200 mm
  • Widely adjustable, elasticated fabric band
  • Conductive fibres on the inside
  • Desco logo moulded into the cap
  • 4 mm stud
Certifications: EN 61340-5-1, ANSI/ESD S1.1.