Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnifiers
Lens, varioLED
Lens, varioLED
Lens, varioLED
Catalog # 630-1221
Supplier:
ESCHENBACH OPTIK
Expand image
Lens, varioLED
Catalog # 630-1221
Supplier:
ESCHENBACH OPTIK
Specifications
Ø:
132 mm
Magnification:
2,5×
Description:
Extra aspheric PXM® lightweight lens, Ceratec® coating for varioLED™
Pk:
1
Specifications
About this item
Product Details
Product Details