



Crystal violet, Sigma-Aldrich®
Catalog # SIAL32675-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:407.99 g/mol
Merck Index:13,04406
Melting Pt:189…194 °C
Density:1.19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-953-6
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS08
:GHS07
:GHS09
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750