ContactUsLinkComponent
Crystal violet, pure, high purity biological stain
Irritant
Health
Corrosive
Environment
Crystal violet, pure, high purity biological stain
Certificates
undefined
Irritant
Health
Corrosive
Environment
Crystal violet, pure, high purity biological stain
Specification Test Results
  • Appearance:
    Dark green to green-brown Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤9 % (110°C)
  • Sulfated ash:
    ≤3 %
  • Titration:
    ≥96.0 % (on dried substance)
  • Lead (Pb):
    ≤30 ppm
  • Zinc (Zn):
    Passes test
  • Insoluble matter:
    (in ethanol) Passes test