Orders LinkContactUsLinkComponent
Silane-Prep Slides
Silane-Prep Slides
Silane prep slides, Glass
Catalog # S4651-72EA
Silane-Prep Slides
Catalog # S4651-72EA
Silane-Prep Slides
Silane prep slides, Glass
Price
115,00 €
Log in for availability
1 * 72 ST
Product Details & Documents
Silane-prep slides are microscope slides coated with silane (aminoalkysilane) to improve the adhesion of tissues to the slide for immunohistochemistry. The slide includes a 'frosted end' for slide/patient identification. It is not tested for use in microarray assays.
  • Silane-prep slides have been used in
  • Immunofluorescence microscopy of DNA fibers
  • Immunohistochemistry
  • The determination of hybridisation stringency
  • Hematology and histology
  • Biological source: synthetic
Specifications
  • Description:
    Silane prep slides
  • Material:
    Glass