Order Entry
Germany
ContactUsLinkComponent
Rhodamine 123 fluorescent dye, mitochondria dye
Rhodamine 123 fluorescent dye, mitochondria dye
Certificates
undefined
Rhodamine 123 fluorescent dye, mitochondria dye

Specification Test Results

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon (dry basis):
    62.2 - 70.2 %
  • Nitrogen (dry basis):
    6.9 - 7.8 %
  • Water (by Karl Fischer):
    ≤10.0 %
  • Solubility (Turbidity)
    1mg/ml in Ethanol:
    Clear to Slightly Hazy
  • Solubility (Colour):
    Conforms to Requirements
    Yellow-Orange
  • Recommended Retest Period:
    4 Years

Specification Test Results