Order Entry
Germany
ContactUsLinkComponent
WypAll® 7041 Rolled Hand Towel Dispenser
WypAll® 7041 Rolled Hand Towel Dispenser
Catalog # 115-2179
Supplier:  KIMBERLY CLARK
undefined
WypAll® 7041 Rolled Hand Towel Dispenser
Catalog # 115-2179
Supplier:  KIMBERLY CLARK

Specifications

  • Colour:
    White
  • Description:
    WypAll® Rolled hand towel plastic dispenser, 7041, 25 cm
  • W×D×H:
    15×15,7×32,4 mm
  • Pk:
    1

Specifications

About this item

The essential rolled dispenser for any business. Compatible with a range of washroom products, this dispenser promotes hygiene and helps to manage costs through controlled dispensing.

  • No touch dispensing, helps reduce contamination and the spread of germs
  • Can be wall mounted for convenience
  • Designed to help increase productivity and reduce waste - practical and easy Clean design with a white, plastic finish with no dirt or dust traps

The practical and easy to clean design makes it the perfect product for any washroom environment. Compatible with Scott® hand towels and WypAll® wipers.